Learn More
Medchemexpress LLC 2-Furoic acid-d3 | 40073-83-4 | 99.99% | 115.10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW012946S50MG
Encompass_Preferred
2-Furoic acid-d3 is the deuterium labeled 2-Furoic acid. 2-Furoic acid (Furan-2-carboxylic acid) is an organic compound produced through furfural oxidation. 2-Furoic acid exhibits hypolipidemic effect, lowering both serum cholesterol and serum triglyceride levels in rats. This compound is for research use only and not sold to patients. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled 2-Furoic acid
  • Organic compound produced through furfural oxidation
  • Exhibits hypolipidemic effect
  • Lowers both serum cholesterol and serum triglyceride levels in rats
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-52138