Learn More
Medchemexpress LLC 2-Furoylglycine | 5657-19-2 | MFCD00030606 | 99.9% | 169.13 g/mol | C7H7NO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1133405MG
Encompass_Preferred
2-Furoylglycine is an endogenous urinary metabolite used as an analytical standard and a putative biomarker for coffee consumption. Supplied as a small-mass research reagent, it is intended for metabolomics, biomarker validation, and analytical reference applications.
  • High purity (99.93%) suitable for analytical work.
  • Small pack sizes for trace-level and reference standard uses.
  • Compatible with common solvents for LC-MS and GC-MS workflows.
  • Molecular formula C7H7NO4; molecular weight 169.13 g/mol.
  • Supplied with product data sheet and safety documentation.
Catalog No. 50-002-64437