Learn More
Apexbio Technology LLC β-Glycerophosphate (sodium salt hydrate)(Synonyms: Sodium β-glycerophosphate hydrate, Sodium glycerophosphate hydrate, Sodium β-glycerophosphate), 10g, CAS: 13408-09-8.
SDP
Supplier:  Apexbio Technology LLC C434710000
Encompass_Preferred
-Glycerophosphate (sodium salt hydrate CAS 13408-09-8) is a biochemical agent commonly employed as a protein phosphatase inhibitor and phosphate donor in cellular research By donating phosphate groups upon hydrolysis in cell culture -glycerophosphate elevates local inorganic phosphate concentrations thereby promoting mineralization It facilitates extracellular matrix calcification in osteoblast models (e g MC3T3-E1 ROS 17/2 8) without altering glycolytic activity Additionally -glycerophosphate in combination with ascorbic acid supports bone matrix mineralization induced by bone morphogenetic protein-2 (BMP-2) in human bone-derived cells and accentuates calcification mediated via alkaline phosphatase activity in cultured vascular smooth muscle cells
Catalog No. 50-000-61116