Learn More
Medchemexpress LLC 2-heptanol, 6-amino-2-methyl- | 372-66-7 | MFCD00127893 | ≥98.0% | 145.25 g/mol | C8H19NO | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW685943100MG
Encompass_Preferred
Heptaminol is an amino alcohol research compound with pressor properties that acts on adrenergic receptors and is used in studies of cardiovascular and neuronal signaling. It is provided for research use and should be stored protected from light at 4°C; in solution store at -80°C for long-term or -20°C short-term.
Catalog No. 50-002-84817