Learn More
TARGETMOL CHEMICALS INC 2-heptyl-3-hydroxy-41H-Quinolo
SDP
Supplier:  TARGETMOL CHEMICALS INC T3837325MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. 2-heptyl-3-hydroxy-4(1H)-Quinolone (Pseudomonas Quinolone Signal) is a quorum-sensing signalling molecule produced by Pseudomonas aeruginosa in response to increased cell density. It increases the expression of the lasB gene in P. aeruginosa, the secretion of the metabolites pyocyanin and the lectin PA-IL, as well as increasing biofilm production in P. aeruginosa populations. It also reduces the iron content of the P. aeruginosa growth medium when used at a concentration of 40 uM and acts as an iron chelator in iron sulphate solutions. Purity 98.06%
Catalog No. 50-287-8984