Learn More
Medchemexpress LLC 2-Heptyl-4-quinolone | 40522-46-1 | 99.1% | 10 MG

Supplier: Medchemexpress LLC HY12950310MG
2-Heptyl-4-quinolone is an intermediate in the synthesis of the Pseudomonas quinolone signal (PQS). It controls swarming by positively regulating phenazine production and induces the production of phenazine-1-carboxylic acid (PCA). This product is intended for research use only.
- Controls swarming by positively regulating phenazine production
- Induces the production of phenazine-1-carboxylic acid (PCA)
- Appears as a solid
- Light yellow to yellow color
- Molecular weight: 243.34
- Formula: C16H21NO
- Classified as an alkaloid, specifically a quinoline alkaloid
- Initially sourced from microorganisms, Pseudomonas aeruginosa
- Storage (solid): Room temperature, protect from light
- Storage (in solvent): -80°C for 1 year, -20°C for 6 months
- Solubility (in vitro): 50 mg/mL in DMSO (205.47 mM); requires ultrasonication
- Exhibits antiproliferative activity with an IC50 value of 35 μM in A2780 cell line
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.