Learn More
Medchemexpress LLC 2-Hexyldecanoic acid 10mM 1mL
SDP
Supplier:  Medchemexpress LLC HYW09963210MM/1ML
Encompass_Preferred
2-Hexyldecanoic acid is a lipid containing one carboxylic acid group and two non-polar tails In the presence of EDC HATU and DCC carboxylic acid groups can undergo continuous amide coupling reactions with amine-containing biomolecules to form stable amide bonds 2-Hexyldecanoic acid can be used as an aerolysin inhibitor to prevent bacterial pathogens 2-Hexyldecanoic acid can cause contact dermatitis[1][2][3]
Catalog No. 50-002-06868