Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydrazinobenzothiazole | 615-21-4 | 99.95% | C7H7N3S | 100 G
SDP

Supplier:  Medchemexpress LLC HY78699100G

Encompass_Preferred

This compound, 2-Hydrazinobenzothiazole, is a potent IDO1 inhibitor with an IC50 of 8 μM. It interacts with the IDO1 haem through hydrazine and has potential for immune system related diseases research. It is supplied with a purity of 99.95%.

  • Potent IDO1 inhibitor (IC50 of 8 μM)
  • Interacts with IDO1 haem via hydrazine
  • Suitable for immune system related diseases research
  • Purity of 99.95%
  • Solid appearance
  • Molecular weight: 165.22
  • Chemical formula: C7H7N3S
  • CAS number: 615-21-4

Catalog No. 50-004-34864


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.