Learn More
eMolecules​ 2-Hydrazinyl-6-methylpyridine | Combi-Blocks | 5315-24-2 | MFCD07786311 | 123.159 | C6H9N3 | 96.000 | Cc1cccc(NN)n1 | 1g | 205398217
Supplier:  eMolecules​ 205398217
Encompass_Preferred
2-Hydrazinyl-6-methylpyridine | Combi-Blocks | 5315-24-2 | MFCD07786311 | 123.159 | C6H9N3 | 96.000 | Cc1cccc(NN)n1 | 1g | 205398217
Catalog No. NC1624650