Learn More
Medchemexpress LLC 2-hydroxy-1-naphthaldehyde | 708-06-5 | MFCD00004005 | 99.9% | 172.18 g/mol | C11H8O2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW016889
Encompass_Preferred
CFL-137 is a small-molecule inhibitor selective for the KRas G12C mutant, used in preclinical oncology research. It exhibits antiproliferative activity across multiple cancer cell lines and reduces tumor growth in mouse models, and is supplied as a high-purity research reagent.
  • Potent KRas G12C inhibitor
  • Shows antiproliferative activity across multiple cancer cell lines
  • Reduces tumor growth in in vivo lung cancer models
  • High purity suitable for research use
  • Soluble in DMSO at ≥100 mg/mL
  • Stable as powder at 4°C; in solution store at -80°C for long-term storage
Catalog No. 50-003-03100