Learn More
Medchemexpress LLC 2-hydroxy-2-methyl-N-(2-(2-(pyridin-3-yloxy)acetyl)-tetrahydroisoquinolin-6-yl)propane-1-sulfonamide | 1698878-14-6 | MFCD28963946 | 99.5% | C20H25N3O5S | 10MG
SDP
Supplier:  Medchemexpress LLC HY1297110MG
Encompass_Preferred
Nampt-IN-1 (LSN3154567) is a potent, selective inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) used in research to study NAD+ biosynthesis, metabolism, and anticancer mechanisms. It inhibits purified NAMPT with an IC50 of 3.1 nM and has been characterized in vitro and in vivo for pharmacokinetics and biological activity.
  • Potent, selective NAMPT inhibition (IC50 = 3.1 nM).
  • Characterized in vitro and in vivo for pharmacokinetics and activity.
  • Useful for cell-based assays probing NAD+ metabolism and proliferation.
  • Supplied as a high-purity research-grade powder suitable for biochemical studies.
  • Documented oral exposure and toxicity profiles in animal models for translational research.
Catalog No. 50-002-59290