Learn More
Medchemexpress LLC 2-hydroxy-3-[(1R)-1-(4-isopropyl-2-furyl)propylamino-cyclobutenylamino]-N,N-dimethylbenzamide | 473728-58-4 | 99.9% | 425.48 g·mol-1 | C23H27N3O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10011100MG
Encompass_Preferred
SCH 563705 is a potent, orally active small-molecule antagonist of the CXC chemokine receptors CXCR2 and CXCR1 used as a research compound to probe receptor signaling in inflammation and oncology. It demonstrates low-nanomolar receptor potency and is supplied as a high-purity solid suitable for in vitro and preclinical in vivo studies.
  • Dual CXCR1/CXCR2 antagonism providing broad chemokine receptor coverage.
  • Low-nanomolar potency (CXCR2 IC50 = 1.3 nM; CXCR1 IC50 = 7.3 nM).
  • High chemical purity for reliable experimental results.
  • Solid form suitable for formulation and dosing in preclinical studies.
  • Characterized by CAS number and molecular weight for accurate identification.
Catalog No. 50-002-15192