Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzaldehyde | 673-22-3 | 152.15 | 100 G
SDP
Supplier:  Medchemexpress LLC HYN0445100G
Encompass_Preferred
2-Hydroxy-4-methoxybenzaldehyde is a potent tyrosinase inhibitor, used in research for its ability to inhibit the oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA). It is also an isomer of Vanillin and can be utilized in the synthesis of Urolithin M7. This compound is a valuable tool for studies related to enzymatic inhibition and organic synthesis.
  • Potent tyrosinase inhibitor.
  • Inhibits L-DOPA oxidation by mushroom tyrosinase.
  • Isomer of Vanillin.
  • Useful in synthesizing Urolithin M7.
Catalog No. 50-003-89897