Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxy-4-methoxybenzaldehyde | 673-22-3 | 152.15 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYN0445

Encompass_Preferred

2-Hydroxy-4-methoxybenzaldehyde is a potent tyrosinase inhibitor and an isomer of Vanillin. It can be used to synthesize Urolithin M7 and inhibits the oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA) by mushroom tyrosinase with an ID50 of 4.3 μg/mL (0.03 mM). It appears as an off-white to light brown solid, being a solid below 41°C and liquid above 43°C.

  • Potent tyrosinase inhibitor
  • Isomer of vanillin
  • Synthesizes urolithin M7
  • Inhibits L-DOPA oxidation
  • Off-white to light brown color
  • Solid below 41°C, liquid above 43°C
  • Molecular weight of 152.15
  • Chemical formula C8H8O3

Catalog No. 50-003-89615


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.