Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzaldehyde | 673-22-3 | 152.15 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYN0445
Encompass_Preferred
2-Hydroxy-4-methoxybenzaldehyde is a potent tyrosinase inhibitor and an isomer of Vanillin. It can be used in the synthesis of Urolithin M7 and inhibits the oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA) by mushroom tyrosinase with an ID50 of 4.3 μg/mL (0.03 mM).
  • Potent tyrosinase inhibitor
  • Isomer of Vanillin
  • Used in the synthesis of Urolithin M7
  • Inhibits L-DOPA oxidation by mushroom tyrosinase
Catalog No. 50-003-89415