Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzaldehyde | 673-22-3 | 152.15 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN044550G
Encompass_Preferred
2-Hydroxy-4-methoxybenzaldehyde is a potent tyrosinase inhibitor and an isomer of Vanillin. It can be used in the synthesis of Urolithin M7. This product is intended for research use only.
  • Inhibits the oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA) by mushroom tyrosinase
  • Can be used to synthesize Urolithin M7
Catalog No. 50-003-88903