Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzaldehyde | 673-22-3 | MFCD00003327 | 152.15 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN04455G
Encompass_Preferred
2-Hydroxy-4-methoxybenzaldehyde is identified as a potent tyrosinase inhibitor and an isomer of Vanillin. This compound has applications in the synthesis of Urolithin M7.
  • Potent tyrosinase inhibitor
  • Isomer of Vanillin
  • Used to synthesize Urolithin M7
  • Inhibits L-DOPA oxidation by mushroom tyrosinase with an ID50 of 4.3 μg/mL
Catalog No. 50-003-88904