Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzoic acid (4-Methoxysalicylic Acid) | 2237-36-7 | 99.6% | C8H8O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7562510MM/1ML
Encompass_Preferred
2-Hydroxy-4-methoxybenzoic acid (4-Methoxysalicylic Acid) is a phenolic compound isolated from *Hemidesmus indicus* and other plants. It exhibits anti-diabetic, hypolipidemic, hepatoprotective, and anti-snake venom activities. This compound can be utilized in the study of diabetes and its complications, liver diseases, immune-related diseases, and snake venom poisoning.
  • Isolated from *Hemidesmus indicus* and other plants
  • Exhibits anti-diabetic, hypolipidemic, hepatoprotective, and anti-snake venom activities
  • Can be utilized in the study of diabetes and its complications
  • Can be utilized in the study of liver diseases
  • Can be utilized in the study of immune-related diseases
  • Can be utilized in the study of snake venom poisoning
  • Has a CC50 of > 40 μM against RAW264.7 cells
  • Soluble in DMSO (50 mg/mL)
Catalog No. 50-004-33754