Learn More
Medchemexpress LLC 2-Hydroxy-4-methylth 250g | 583-91-5 | 150.20 g/mol | C5H10O3S | 250 G

Supplier: Medchemexpress LLC HY116688250G
2-Hydroxy-4-(methylthio)butyric acid is a sulfur-containing hydroxybutyric acid used as a reagent in synthetic chemistry and in studies of amino acid metabolism. It is supplied as a liquid and is available in laboratory package sizes, including 250 G.
- Hydroxy analogue of methionine used in synthetic and metabolic studies.
- Supplied as a light yellow to brown liquid.
- Molecular formula C5H10O3S; molecular weight 150.20 g/mol.
- Purity (NMR) 88.0% as reported on the COA.
- Available in multiple package sizes, including 250 G.
- Certificate of analysis and safety data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.