Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Hydroxy-5'-methoxyacetophenone | 705-15-7 | 166.18 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYN173110MM/1ML

Encompass_Preferred

2'-Hydroxy-5'-methoxyacetophenone is an acetophenone derivative with acaricidal activities. It attenuates the inflammatory response via the NF-κB signaling pathway. It also exhibits significant inhibitory activity against α-amylase, collagenase, and aldose reductase (AR), suggesting its potential in combating diabetes. Additionally, it exhibits anti-ovarian cancer activity.

  • Purity: 99.67% or 99.96%
  • Appearance: Solid
  • Color: Light yellow to green yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (601.76 mM)

Catalog No. 50-004-16392


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.