Learn More
Medchemexpress LLC 2-hydroxy-5-nitrobenzoic acid | 96-97-9 | MFCD00007338 | 96.8% | 183.12 g/mol | C7H5NO5 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01635250G
Encompass_Preferred
2-hydroxy-5-nitrobenzoic acid is a nitrosalicylic acid derivative used as an HPLC analytical standard, a pH indicator, and a reagent for heavy-metal detection. It is also used in research on the oxidation of phenolic compounds and other analytical chemistry applications.
  • Used as an HPLC analytical standard.
  • Functions as a pH indicator for analytical assays.
  • Serves as a reagent for heavy-metal detection.
  • Applicable to studies of phenolic compound oxidation.
  • Supplied with documented purity and certificate of analysis for analytical use.
Catalog No. 50-003-02079