Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hydroxy-6-methoxybenzoic acid | 3147-64-6 | MFCD00674090 | 99.3% | 168.15 g/mol | C8H8O4 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYW01710010MM/1ML

Encompass_Preferred

2-Hydroxy-6-methoxybenzoic acid is a small-molecule research chemical used in biochemical studies and organic synthesis. It is provided as a solid and as ready-to-use DMSO solutions for convenient handling, rapid experimental setup, and reliable analytical characterization.

  • High stated purity (99.34%).
  • Molecular formula C8H8O4; molecular weight 168.15 g/mol.
  • Available as solid and 10 mM solution in DMSO (1 mL).
  • CAS number 3147-64-6 for unequivocal identification.
  • Storage guidance: powder at -20°C (long term) or 4°C; in solvent at -80°C recommended.
  • Documents available: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-003-00338


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.