Learn More
Medchemexpress LLC 2-hydroxy-6-methoxybenzoic acid | 3147-64-6 | MFCD00674090 | 99.3% | 168.15 g/mol | C8H8O4 | 10 G

Supplier: Medchemexpress LLC HYW01710010G
2-hydroxy-6-methoxybenzoic acid (6-methoxysalicylic acid) is a substituted benzoic acid used as a research reagent and synthetic intermediate in organic and medicinal chemistry. It is identified by CAS 3147-64-6 and is commonly supplied as a solid for laboratory use.
- High purity (99.3%) determined by HPLC.
- Molecular formula C8H8O4.
- Molecular weight 168.15 g/mol.
- Suitable as a synthetic intermediate and analytical standard.
- Provided with COA and analytical data for batch verification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.