Learn More
Cayman Chemical 2-hydroxy FlutamIde 10mg
SDP
Supplier:  Cayman Chemical 1527110 MG
Encompass_Preferred
The major metabolite formed during the metabolism of the non-steroidal antiandrogen flutamide by cytochrome P450 isoforms CYP1A2 and CYP3A4; inhibits the binding of testosterone to the nuclear androgen receptor (IC50 = ~300-900), blocking the expression of AR target genes
Catalog No. 50-297-2843