Learn More
Medchemexpress LLC 2-Hydroxyacetophenone | 582-24-1 | 136.15 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW002198100G
Encompass_Preferred
2-Hydroxyacetophenone is a chemical compound that serves as a principal root volatile of *Carissa edulis* and can be used as a versatile building block. It appears as an off-white to light yellow solid.
  • Inhibits infection of HIV/SARS-CoV S pseudovirus.
  • Acts as a substrate for *Lactobacillus brevis* alcohol dehydrogenase (LBADH).
  • Can be used as a building block.
  • Off-white to light yellow solid appearance.
Catalog No. 50-004-46493