Learn More
Medchemexpress LLC 2-Hydroxyacetophenone | 582-24-1 | 136.15 | 50 G
Supplier: Medchemexpress LLC HYW00219850G
2-Hydroxyacetophenone, also known as Phenacyl alcohol, is identified as a principal root volatile of the Carissa edulis plant. It serves as a versatile building block for various applications. The compound has demonstrated inhibitory effects on HIV/SARS-CoV S pseudovirus infection and functions as a substrate for Lactobacillus brevis alcohol dehydrogenase (LBADH).
- Principal root volatile of Carissa edulis
- Building block for various applications
- Inhibitory effects on HIV/SARS-CoV S pseudovirus infection
- Substrate for Lactobacillus brevis alcohol dehydrogenase