Learn More
Medchemexpress LLC 2-Hydroxybenzimidazole | 615-16-7 | 134.14 | 50 G
Supplier: Medchemexpress LLC HYW00800350G
2-Hydroxybenzimidazole is an inactive compound targeting CYP1A1. It is mainly used as a control compound in the structure-activity relationship study of benzimidazole derivatives. It does not induce CYP1A1 expression like its thiol or amino substituted derivatives. This product is for research use only and not for sale to patients.
- Targets CYP1A1 as an inactive compound
- Used as a control in benzimidazole derivative studies
- Does not induce CYP1A1 expression
- For research use only