Learn More
Selleck Chemical LLC 2-Hydroxybenzyl alcohol
SDP
Supplier:  Selleck Chemical LLC S556225MG
Encompass_Preferred
2-Hydroxybenzyl alcohol (Salicyl alcohol Salicain Diathesin Saligenin Saligenol o-Methylolphenol 2-Toluenediol) which can be found in a number of food items such as red huckleberry rye jerusalem artichoke and ceylon cinnamon is precursor of salicylic acid and is formed from salicin by enzymatic hydrolysis by Salicyl-alcohol beta-D-glucosyltransferase or by acid hydrolysis
Catalog No. 50-000-03814