Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxydecanoic ac 100mg | 5393-81-7 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY114757100MG

Encompass_Preferred

2-Hydroxydecanoic acid is a medium-chain α-hydroxy fatty acid used as a biochemical reagent in life-science research. Supplied as a small, ready-to-use solid, it is intended for biochemical assays and studies that require 2-hydroxy fatty acids.

  • Purity 98.0%.
  • Molecular formula C10H20O3.
  • Molecular weight 188.26 g·mol⁻1.
  • Provided as a 100 mg quantity.
  • Suitable for biochemical assays and reagent applications.

Catalog No. 50-002-28998


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.