Learn More
Medchemexpress LLC 2-Hydroxydecanoic ac 25mg | 5393-81-7 | 25 MG

Supplier: Medchemexpress LLC HY11475725MG
2-Hydroxydecanoic acid is a racemic α-hydroxy medium-chain fatty acid supplied as a solid reagent for biochemical and lipid research. Provided in small weighed packs, it is suitable for analytical assays, method development, and preparative workflows.
- Purity 98.0% for analytical and experimental use.
- Available in small, convenient 25 mg packs for accurate dosing.
- Solid appearance simplifies handling and storage.
- Storage: stable at room temperature for long-term storage; in solution store at -80 °C or -20 °C per guidelines.
- Molecular weight 188.26 g/mol; formula C10H20O3.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.