Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxydecanoic ac 50mg | 5393-81-7 | 188.26 g/mol | C10H20O3 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY11475750MG

Encompass_Preferred

2-Hydroxydecanoic acid (±)-2-hydroxydecanoic acid is an α-hydroxy medium-chain fatty acid used as a biochemical reagent for life-science research. It is supplied in a small-scale vial for assay and analytical applications.

  • High purity: 98.0%.
  • Molecular formula C10H20O3.
  • Molecular weight 188.26 g/mol.
  • CAS number 5393-81-7.
  • Pack size: 50 MG vial suitable for analytical and biochemical assays.

Catalog No. 50-002-29715


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.