Learn More
Medchemexpress LLC 2-Hydroxyethyl methacrylate | 868-77-9 | 99.1% | 130.14 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW012723500G
Encompass_Preferred
2-Hydroxyethyl methacrylate (HEMA) is a monomer used in the synthesis of various polymers. Its polymer, PHEMA, is extensively employed in the creation of dental composite materials.
  • High purity of 99.05%
  • Liquid appearance
  • Colorless to light yellow
  • Specific molecular formula C6H10O3
  • CAS number 868-77-9
Catalog No. 50-004-54793