Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxymethyl-5-hydroxypyridine | 40222-77-3 | 125.13 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYW00714010MM/1ML

Encompass_Preferred

2-Hydroxymethyl-5-hydroxypyridine is isolated from the matured, ripened and dried seeds of S. lychnophora. It is supplied as a 1 mL solution with a concentration of 10 mM in DMSO, having a high purity of 99.7% and a molecular weight of 125.13. This product is intended for research purposes only.

  • Isolated from S. lychnophora seeds
  • Purity of 99.7%
  • Molecular weight of 125.13
  • Supplied as a 10 mM solution in DMSO
  • Provided in a 1 mL volume
  • Store at -80°C for 6 months or -20°C for 1 month when in solvent
  • For research use only

Catalog No. 50-004-60256


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.