Learn More
Medchemexpress LLC 2-Hydroxypalmitic acid | 764-67-0 | 1 G
SDP
Supplier:  Medchemexpress LLC HYN78141G
Encompass_Preferred
2-Hydroxypalmitic acid is an intermediate in the phytosphingosine metabolic pathway. It is exclusively incorporated into sphingolipids and not utilized for other lipids. This compound promotes the production of odd-carbon phosphatidylcholine species and is applicable to studies of Saccharomyces cerevisiae infection. It is suitable for laboratory research only.
  • Intermediate in the phytosphingosine metabolic pathway
  • Exclusively incorporated into sphingolipids
  • Promotes production of odd-carbon phosphatidylcholine species
  • Applicable to studies of Saccharomyces cerevisiae infection
  • Suitable for research use only
Catalog No. 50-004-19085