Learn More
Medchemexpress LLC 2-Hydroxypalmitic acid | 764-67-0 | MFCD00014343 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN7814500MG
Encompass_Preferred
2-Hydroxypalmitic acid is an intermediate within the phytosphingosine metabolic pathway, exclusively incorporated into sphingolipids rather than other lipids. This compound promotes the production of odd-carbon phosphatidylcholine species and is applicable for studies concerning *Saccharomyces cerevisiae* infection.
  • Intermediate in phytosphingosine metabolic pathway
  • Exclusively incorporated into sphingolipids
  • Promotes production of odd-carbon phosphatidylcholine species
  • Applicable for studies concerning *Saccharomyces cerevisiae* infection
Catalog No. 50-004-17703