Learn More
Medchemexpress LLC 2-Hydroxyquinoline | 59-31-4 | 145.16 | 1 ML

Supplier: Medchemexpress LLC HYW00559810MM/1ML
2-Hydroxyquinoline is an off-white to light yellow solid. It acts as an inhibitor of α-glucosidase and α-amylase, with IC50 values of 64.4 µg/mL and 130.5 µg/mL respectively. This product is suitable for research in the study of diabetes.
- Inhibits α-glucosidase and α-amylase
- Targeted to glycosidase and amylases
- In vitro solubility in DMSO at 100 mg/mL
- Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
- Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
- Purity by HPLC is 99.4%
- 1H NMR spectrum consistent with structure
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.