Learn More
Medchemexpress LLC 2-Iodoaniline | 615-43-0 | MFCD00007680 | 219.03 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00731810MM/1ML
Encompass_Preferred
2-Iodoaniline (2-Iodophenylamine) is an aniline derivative with potential hepatotoxic and nephrotoxic activity, intended for research use only.
  • Aniline derivative
  • Exhibits potential hepatotoxic activity
  • Exhibits potential nephrotoxic activity
  • Induces oliguria, diminished kidney weight, and tubular casts in animal models
  • Decreases renal cortical slice accumulation of organic anions
  • Elevates plasma ALT/GPT activity and alters morphology in the centrilobular region
  • For research use only
Catalog No. 50-004-60348