Learn More
Cayman Chemical 2-IodomelatonIn 10mg
SDP
Supplier:  Cayman Chemical 1971110 MG
Encompass_Preferred
A potent MT1 receptor agonist (Ki = 28 pM); 5-fold selective for MT1 over MT2; inhibits forskolin-stimulated cAMP production in CHO cells expressing human MT1 (EC50 = 11 pM); has been used to characterize the role of MT1 in melatonin-mediated signaling
Catalog No. 50-298-2339