Learn More
Medchemexpress LLC (2-Iodophenyl)methanol | 5159-41-1 | 234.03 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00467810G
Encompass_Preferred
(2-Iodophenyl)methanol is a biochemical reagent intended for life science research. It serves as a versatile biological material or organic compound for various applications. This product is strictly for research purposes and not for patient use.
  • Biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Used in the synthesis of substituted seven-membered lactones
  • Employed in the synthesis of 2-[(E)-(1 -iodo-2 -propenyl)]benzyl alcohol
  • Utilized in the synthesis of 2,3-diphenyl-1-indenone
Catalog No. 50-004-49066