Learn More
Medchemexpress LLC (2-Iodophenyl)methanol | 5159-41-1 | 234.03 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW004678
Encompass_Preferred
(2-Iodophenyl)methanol is a biochemical reagent that appears as a white to off-white solid. It has a molecular weight of 234.03 and the chemical formula C7H7IO. This compound is suitable for life science research and has been used in the synthesis of substituted seven-membered lactones and various benzyl alcohol derivatives.
  • High purity at 99.81%
  • Can be used as a biological material or organic compound
  • Suitable for life science-related research
  • Used in the synthesis of substituted seven-membered lactones and benzyl alcohol derivatives
Catalog No. 50-004-49186