Learn More
Medchemexpress LLC 2-isobutyl-3-methoxypyrazine-d3 | 588732-63-2 | 98.0% | 169.24 g/mol | C9H11D3N2O | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW017141S1MG
Encompass_Preferred
2-Isobutyl-3-methoxypyrazine-d3 is the deuterium-labeled analogue of 2-isobutyl-3-methoxypyrazine supplied as a stable isotope reference standard for analytical use. It is suited for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS workflows for analysis of aroma compounds in food, beverage, and biological samples.
  • Deuterium-labeled analogue for isotope dilution and tracer studies.
  • Compatible with NMR, GC-MS, and LC-MS quantitation workflows.
  • Facilitates accurate quantitation of trace aroma compounds.
  • Offered in small, pre-weighed quantities for analytical use.
  • Supplied in pure form with recommended storage conditions.
Catalog No. 50-003-00898