Learn More
Research Products International Corp 2-Keto Acid Mono Salt 500 G

Supplier: Research Products International Corp K92100500.0
CAS Number: 997-43-3
Molecular Formula: C5H5O5K
Molecular Weight: 184.19
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.