Learn More
US Plastic Corporation 2 LITER STR CLEAR ERL FLASK 2L
SDP
Supplier:  US Plastic Corporation 78952
Encompass
2 Liter Sterile Clear Erlenmeyer Flasks with White 53mm VersaCap. Case/6. Non Returnable/Non Cancel
Catalog No. NC1953618