Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-maleimido acetic acid | 25021-08-3 | MFCD00457254 | 100.0% | 155.11 g/mol | C6H5NO4 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW017831100MG

Encompass_Preferred

2-Maleimido acetic acid is a carboxylic acid bearing an active maleimide functional group used as an intermediate in organic synthesis and bioconjugation. It is supplied as a solid and is commonly prepared as DMSO solutions for downstream coupling reactions.

  • Contains a reactive maleimide moiety for thiol-selective coupling.
  • Suitable as an intermediate for synthesis of linkers and functionalized molecules.
  • High purity for reliable experimental results.
  • Soluble in DMSO; ready-to-use solutions are available.
  • Provided in lab-scale pack sizes for research workflows.

Catalog No. 50-003-00619


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.