Learn More
Medchemexpress LLC 2-mercaptobenzothiazole | 149-30-4 | MFCD00005781 | 99.5% | 167.25 | C7H5NS2 | 100 G

Supplier: Medchemexpress LLC HYW017113100G
2-Mercaptobenzothiazole is a sulfur-containing benzothiazole used as a chemical reagent and analytical standard for research applications. It is reported to activate the aryl hydrocarbon receptor and to inhibit thyroid hormone synthesis and dopamine β-hydroxylase.
- CAS 149-30-4 for unambiguous identification.
- Molecular formula C7H5NS2 and molecular weight 167.25.
- Solid, white to yellow appearance suitable for storage and handling.
- High reported purity (~99.5%) with downloadable COA and SDS available.
- Available in multiple pack sizes, including 100 g, 500 g, and 1 kg.
- Commonly used as an analytical standard and research reagent in biochemical assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.