Learn More
Supply Solutions 2-Mercaptoethanol, 60-24-2, MFCD00004890, 25mL
Supplier:  Supply Solutions SIGMA M314825ML
Encompass_Preferred
Linear Formula HSCH2CH2OH, 99% (GC/titration), Molecular Weight 78.13, bp 157 C (lit.), for molecular biology, for electrophoresis, suitable for cell culture, BioReagent, Synonym: ?-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol
Catalog No. 50-161-3343