Learn More
ILEX LIFE SCIENCES LLC 2-MERCAPTOETHANOL 50MM 6X100ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0705100X6
Encompass_Preferred
PAN-Biotech 2-Mercaptoethanol 50 mM in DPBS (6 x 100 ml). DESCRIPTION: 2-Mercaptoethanol, also known as beta-mercaptoethanol (BME), is a potent reducing agent used in cell culture medium to prevent toxic levels of oxygen radicals. 2-Mercaptoethanol is not stable in solution, thus protocols generally require daily supplementation. PAN-Biotech 2-Mercaptoethanol is supplied at a concentration of 50 mM in Dulbecco's Phosphate Buffered Saline (DPBS). SOLUTION: 50 mM in DPBS. STORAGE: +2-8°C. STERILE: Yes. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany.
Catalog No. 50-273-3715