Learn More
Cayman Chemical 2-Methoxy-4-methylphenol 250g
SDP
Supplier:  Cayman Chemical 36533250 G
Encompass_Preferred
A phenolic pyrolysis product of plant biomass; a volatile component of black olives and roasted coffee beans
Catalog No. 50-291-1601