Learn More
Medchemexpress LLC 2-Methoxybenzoic acid | 579-75-9 | MFCD00002431 | 152.15 | 1 ML

Supplier: Medchemexpress LLC HYN139310MM/1ML
2-Methoxybenzoic acid (NSC 3778) is a human endogenous metabolite used as an internal standard of salicylic acid and its putative biosynthetic precursors in cucumber leaves. It also finds application in the synthesis of Benextramine.
- Purity of 99.86%
- Appears as a white to off-white solid
- Soluble in DMSO at 100 mg/mL
- Powder stable for 3 years at -20°C or 2 years at 4°C
- In solvent, stable for 2 years at -80°C or 1 year at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.