Learn More
Medchemexpress LLC 2-Methoxycarbonylphenylboronic acid | 374538-03-1 | 179.97 | 10 G

Supplier: Medchemexpress LLC HYW00290810G
2-Methoxycarbonylphenylboronic acid is an intermediate for laboratory use in the synthesis of BLT2 agonists and for the manufacture of substances.
- Appears as a white to off-white solid.
- Soluble in DMSO at 200 mg/mL (1111.30 mM), requiring ultrasonic.
- Store powder at -20°C for up to 3 years or 4°C for up to 2 years.
- Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month.
- Shipped at room temperature for durations less than 2 weeks.
- Intended for research use only by experienced personnel.
- Melting/freezing point is 90-96°C.
- Boiling point is 359°C at 760 mmHg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.